accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)N(R)O(S)(R)NNOHNH2HOHOCl

CAS RN 146-77-0

IUPAC Name: (2R,3R,4S,5R)-2-(6-amino-2-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: 2-chloroadenosine
Molecular Weight (g/mol): 301.69
Molecular Formula: C10H12ClN5O4
InChi Key: BIXYYZIIJIXVFW-SBTUYZBDNA-N
SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC(Cl)=N1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)