accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOHNCH3H3C

CAS RN 1465-16-3

IUPAC Name: 3-[2-(dimethylamino)ethyl]-1-methyl-1H-indol-4-ol
Synonyms: 1-methylpsilocin
Molecular Weight (g/mol): 218.3
Molecular Formula: C13H18N2O
InChi Key: MZZRFEIDRWKTKJ-UHFFFAOYSA-N
SMILES: CN(C)CCC1=CN(C)C2=CC=CC(O)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results