accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)NH3OOOO

CAS RN 146669-29-6

IUPAC Name: 4-[(1R)-1-azaniumyl-1-carboxylatoethyl]benzoate
Synonyms: 4-[(1R)-1-ammonio-1-carboxylatoethyl]benzoate
Molecular Weight (g/mol): 208.19
Molecular Formula: C10H10NO4
InChi Key: DNCAZYRLRMTVSF-SNVBAGLBSA-M
SMILES: C[C@]([NH3+])(C([O-])=O)C1=CC=C(C=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)