accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CH3CCH3H3CNSNBOOCH3CH3H3CCH3NSN

CAS RN 1467776-41-5

IUPAC Name: 7,15-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4,12-dithia-3,5,11,13-tetraazatetracyclo[7.7.0.0²,⁶.0¹⁰,¹⁴]hexadeca-1(9),2,5,7,10,13,15-heptaene
Synonyms: 7,15-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4,12-dithia-3,5,11,13-tetraazatetracyclo[7.7.0.0²,⁶.0¹⁰,¹⁴]hexadeca-1(9),2,5,7,10,13,15-heptaene
Molecular Weight (g/mol): 496.21
Molecular Formula: C22H26B2N4O4S2
InChi Key: VZCYKOASVMGCSP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C(B3OC(C)(C)C(C)(C)O3)C3=NSN=C23)C2=NSN=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)