CAS RN 147059-75-4
IUPAC Name:
7-[(1R,5S)-6-amino-3-azabicyclo[3.1.0]hexan-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid; methanesulfonic acid
Synonyms:
methanesulfonic acid; trovafloxacin
Molecular Weight (g/mol):
512.46
Molecular Formula:
C21H19F3N4O6S
InChi Key:
DYNZICQDCVYXFW-GIPYJWDTSA-N
SMILES:
CS(O)(=O)=O.NC1[C@@H]2CN(C[C@H]12)C1=C(F)C=C2C(=O)C(=CN(C3=CC=C(F)C=C3F)C2=N1)C(O)=O
1
–
1
of
1
results
1
Tocris Bioscience™ Trovafloxacin mesylate
CAS: 147059-75-4 Molecular Formula: C21H19F3N4O6S