Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Trovafloxacin mesylate
GREENER_CHOICESDP

Catalog No. 386350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-635-0 50 mg
38-631-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-635-0 Supplier Tocris Bioscience™ Supplier No. 3863/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Antibiotic; inhibits bacterial DNA synthesis

Fluoroquinolone antibiotic. Inhibits bacterial DNA topoisomerase IV and DNA gyrase and forms a stable quinolone-DNA complex with these enzymes which reversibly inhibits DNA synthesis. Displays potent activity against gram-positive and gram-negative bacteria. Increases the production of mitochondrial NO in immortalized hepatocytes; also increases mitochondrial Ca2+. Inhibits Panx-1 (IC50 ∽ 4μM).

Chemical Identifiers

CAS 147059-75-4
Molecular Formula C21H19F3N4O6S

Specifications

Quantity 50 mg
Formula Weight 512.46
Purity >99%
Molecular Weight (g/mol) 512.46
Chemical Name or Material Trovafloxacin mesylate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.