accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3NOSNH3CSNH3C(E/Z)(E)

CAS RN 147366-41-4

IUPAC Name: 1-ethyl-2-({3-ethyl-5-[(2E)-3-methyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]-4-oxo-1,3-thiazolidin-2-ylidene}methyl)pyridin-1-ium chloride
Synonyms: 1-ethyl-2-({3-ethyl-5-[(2E)-3-methyl-1,3-benzothiazol-2-ylidene]-4-oxo-1,3-thiazolidin-2-ylidene}methyl)pyridin-1-ium chloride
Molecular Weight (g/mol): 432
Molecular Formula: C21H22ClN3OS2
InChi Key: VSKYOTRJSLYFHX-UXJRWBAGSA-M
SMILES: [Cl-].CCN1C(S\C(C1=O)=C1\SC2=CC=CC=C2N1C)=CC1=CC=CC=[N+]1CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)