Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MKT 077
SDP

Catalog No. RD10195060 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10195060 10 mg
46-215-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RD10195060 Supplier R&D Systems™ Supplier No. 4621/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Tocris product, part of R&D Systems. Binds mot-2; selectively cytotoxic in cancer cells

Occupies mortalin-2 (mot-2), a member of the Hsp70 family, at its p53 binding site and enables p53 translocation to the nucleus. Selectively cytotoxic; causes growth arrest of cancer cells in culture. Also inhibits telomerase activity and cross-links F-actin.

Chemical Identifiers

CAS 147366-41-4
Molecular Formula C21H22ClN3OS2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 432
Purity >98%
Molecular Weight (g/mol) 432.00
Chemical Name or Material MKT 077
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.