CAS RN 148152-66-3
IUPAC Name:
2-(6-chloropyridin-3-yl)-7-azabicyclo[2.2.1]heptane
Synonyms:
2-(6-chloropyridin-3-yl)-7-azabicyclo[2.2.1]heptane
Molecular Weight (g/mol):
208.69
Molecular Formula:
C11H13ClN2
InChi Key:
NLPRAJRHRHZCQQ-UHFFFAOYNA-N
SMILES:
ClC1=NC=C(C=C1)C1CC2CCC1N2
1
–
1
of
1
results
1
Tocris Bioscience™ (±)-Epibatidine
CAS: 148152-66-3 Molecular Formula: C11H13ClN2