Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (±)-Epibatidine
GREENER_CHOICESDP

Catalog No. 068410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-841-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-841-0 Supplier Tocris Bioscience™ Supplier No. 0684/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity nAChR agonist

High affinity nicotinic agonist (Ki values are 0.02 and 233 nM for α4β2 and α7 nicotinic receptors respectively). Analgesic.

Chemical Identifiers

CAS 148152-66-3
Molecular Formula C11H13ClN2

Specifications

Quantity 10 mg
Formula Weight 208.69
Purity 98%
Molecular Weight (g/mol) 208.689
Chemical Name or Material (±)-Epibatidine
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.