CAS RN 148687-76-7
IUPAC Name:
3-{[3,5-bis(trifluoromethyl)phenyl]methoxy}-2-phenylpiperidine hydrochloride
Synonyms:
3-{[3,5-bis(trifluoromethyl)phenyl]methoxy}-2-phenylpiperidine hydrochloride
Molecular Weight (g/mol):
439.83
Molecular Formula:
C20H20ClF6NO
InChi Key:
DYEUTIUITGHIEO-UHFFFAOYNA-N
SMILES:
Cl.FC(F)(F)C1=CC(=CC(COC2CCCNC2C2=CC=CC=C2)=C1)C(F)(F)F
1
–
1
of
1
results
1
R&D Systems™ L-733,060 hydrochloride
CAS: 148687-76-7 Molecular Formula: C20H20ClF6NO