accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3HNONNOCH3HClHCl

CAS RN 149007-54-5

IUPAC Name: N-tert-butyl-3-[4-(2-methoxyphenyl)piperazin-1-yl]-2-phenylpropanamide dihydrochloride
Synonyms: N-tert-butyl-3-[4-(2-methoxyphenyl)piperazin-1-yl]-2-phenylpropanamide dihydrochloride
Molecular Weight (g/mol): 468.46
Molecular Formula: C24H35Cl2N3O2
InChi Key: VJGZNBYDSDEOED-UHFFFAOYNA-N
SMILES: Cl.Cl.COC1=CC=CC=C1N1CCN(CC(C(=O)NC(C)(C)C)C2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results