accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 149301-79-1

IUPAC Name: (6Z,9Z,12Z,15Z)-1,1,1-trifluorohenicosa-6,9,12,15-tetraen-2-one
Synonyms: AACOCF3 arachidonic acid trifluoromethyl ketone aacocf3 arachidonyl trifluoromethyl ketone arachidonyltrifluoromethane
Molecular Weight (g/mol): 356.47
Molecular Formula: C21H31F3O
InChi Key: PLWROONZUDKYKG-DOFZRALJSA-N
SMILES: CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)