Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AACOCF3
GSA_VAGREENER_CHOICESDP

Catalog No. 14625 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-625 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-625 Supplier Tocris Bioscience™ Supplier No. 1462/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Phospholipase A2 inhibitor

Inhibitor of cytosolic (85 kDa) phospholipase A2. Also inhibits fatty acid amide hydrolase (FAAH, anandamide amidase) in vitro.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 149301-79-1
Molecular Formula C21H31F3O

Specifications

Quantity 5 mg
Formula Weight 356.47
Purity >98%
Molecular Weight (g/mol) 356.47
Chemical Name or Material AACOCF3
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.