accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3SOON(R)ON(S)H2NO(S)HOClCl

CAS RN 150375-75-0

IUPAC Name: (2S)-1-[(2R,3S)-5-chloro-3-(2-chlorophenyl)-1-(3,4-dimethoxybenzenesulfonyl)-3-hydroxy-2,3-dihydro-1H-indole-2-carbonyl]pyrrolidine-2-carboxamide
Synonyms: relcovaptan
Molecular Weight (g/mol): 620.5
Molecular Formula: C28H27Cl2N3O7S
InChi Key: CEBYCSRFKCEUSW-NAYZPBBASA-N
SMILES: COC1=CC=C(C=C1OC)S(=O)(=O)N1[C@@H](C(=O)N2CCC[C@H]2C(N)=O)[C@](O)(C2=CC(Cl)=CC=C12)C1=CC=CC=C1Cl
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results