Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 49059
GREENER_CHOICESDP

Catalog No. 231010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-101-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-101-0 Supplier Tocris Bioscience™ Supplier No. 2310/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective, orally active vasopressin V1A receptor antagonist

Potent and selective non-peptide vasopressin V1A receptor antagonist; devoid of agonist activity. Displays high affinity and efficacy at both rat (Ki = 1.6 nM) and human (Ki = 1.1 - 6.3 nM) V1A receptors. Potently antagonizes arginine vasopressin-induced effects in vitro (IC50 = 3.7 nM for inhibition of human platelet aggregation) and is orally active in vivo.

Specifications

CAS 150375-75-0
Quantity 10 mg
Product Type SR 49059
Purity >99%
Molecular Weight (g/mol) 620.498
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.