CAS RN 150651-39-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ iFSP1
Ferroptosis suppressor protein 1 (FSP1) inhibitor; ferroptosis inducer