Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ iFSP1
GREENER_CHOICESDP

Catalog No. 716250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-625-0 50 mg
71-621-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 71-625-0 Supplier Tocris Bioscience™ Supplier No. 7162/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ferroptosis suppressor protein 1 (FSP1) inhibitor; ferroptosis inducer

iFSP1 is a ferroptosis inducer. Inhibits ferroptosis suppressor protein 1 (FSP1) and induces ferroptosis in GPX4 knockout cells. Sensitizes tumor cells to RSL3 (Cat. No. 6118) -induced ferroptosis.

Specifications

Chemical Name or Material 1-Amino-3-(4-methylphenyl)-pyrido[1,2-a]benzimidazole-2,4-dicarbonitrile
CAS 150651-39-1
Quantity 50 mg
Target Other Reductase Inhibitors
Molecular Formula C20H13N5
Formula Weight 5252
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.