accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OOHNOONNHNNOOHNOONNHNNO

CAS RN 150821-03-7

IUPAC Name: bis(N-[4-oxo-2-(2H-1,2,3,4-tetrazol-5-yl)-4H-chromen-8-yl]-4-(4-phenylbutoxy)benzamide) hydrate
Synonyms: bis(pranlukast) hydrate
Molecular Weight (g/mol): 981.04
Molecular Formula: C54H48N10O9
InChi Key: MSXTUBJFNBZPGC-UHFFFAOYSA-N
SMILES: O.O=C(NC1=C2OC(=CC(=O)C2=CC=C1)C1=NNN=N1)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1.O=C(NC1=C2OC(=CC(=O)C2=CC=C1)C1=NNN=N1)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results