accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 151126-84-0

IUPAC Name: sodium 5-(2-{[2-(3-chlorophenyl)-2-hydroxyethyl]amino}propyl)-2H-1,3-benzodioxole-2,2-dicarboxylate
Synonyms: sodium 5-(2-{[2-(3-chlorophenyl)-2-hydroxyethyl]amino}propyl)-1,3-benzodioxole-2,2-dicarboxylate
Molecular Weight (g/mol): 442.8
Molecular Formula: C20H18ClNNaO7
InChi Key: DBEDHSNRCKDNPA-UHFFFAOYNA-L
SMILES: [Na+].CC(CC1=CC2=C(OC(O2)(C([O-])=O)C([O-])=O)C=C1)NCC(O)C1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results