Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CL 316243 disodium salt
GREENER_CHOICESDP

Catalog No. 149950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-995-0 50 mg
14-991-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-995-0 Supplier Tocris Bioscience™ Supplier No. 1499/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly selective β3 agonist

Potent and highly selective β3-adrenoceptor agonist (EC50 = 3 nM); > 10000-fold selective over β1 and β2 receptors. Increases brown adipose tissue thermogenesis and metabolic rate, and decreases blood insulin and glucose levels following oral administration in vivo.

Chemical Identifiers

CAS 151126-84-0
Molecular Formula C20H18ClNNaO7-

Specifications

Quantity 50 mg
Formula Weight 465.8
Purity >96%
Molecular Weight (g/mol) 442.804
Chemical Name or Material CL 316243 disodium salt
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.