accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOOFFFOSOOFFF

CAS RN 152873-79-5

IUPAC Name: 5-(trifluoromethanesulfonyloxy)naphthalen-1-yl trifluoromethanesulfonate
Synonyms: 5-(trifluoromethanesulfonyloxy)naphthalen-1-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 424.28
Molecular Formula: C12H6F6O6S2
InChi Key: IFHGSECJTKJJIJ-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC=CC2=C(OS(=O)(=O)C(F)(F)F)C=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results