accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2HOOH3CCH3CH3H3C

CAS RN 153559-49-0

IUPAC Name: 4-[1-(3,5,5,8,8-pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl]benzoic acid
Synonyms: bexarotene targrexin bexaroteno bexarotenum targretin-gel targretin targretyn targret
Molecular Weight (g/mol): 348.49
Molecular Formula: C24H28O2
InChi Key: NAVMQTYZDKMPEU-UHFFFAOYSA-N
SMILES: CC1=CC2=C(C=C1C(=C)C1=CC=C(C=C1)C(O)=O)C(C)(C)CCC2(C)C
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)