CAS RN 154039-60-8
IUPAC Name:
(2S,3R)-N'-[(1S)-2,2-dimethyl-1-(methylcarbamoyl)propyl]-N,2-dihydroxy-3-(2-methylpropyl)butanediamide
Synonyms:
marimastat 马立马司他
Molecular Weight (g/mol):
331.41
Molecular Formula:
C15H29N3O5
InChi Key:
OCSMOTCMPXTDND-OUAUKWLOSA-N
SMILES:
CNC(=O)[C@@H](NC(=O)[C@H](CC(C)C)[C@H](O)C(=O)NO)C(C)(C)C
1
–
2
of
2
results
1
Thermo Scientific Chemicals Marimastat, 98%
CAS: 154039-60-8 Molecular Formula: C15H29N3O5 Synonym: (2S,3R)-N4-[(1S)-2,2-Dimethyl-1-[(methylamino)carbonyl] propyl]-N1,2-dihydroxy-3-(2-methylpropyl)butanediamide; BB2516
2
R&D Systems™ Marimastat
CAS: 154039-60-8 Molecular Formula: C15H29N3O5