accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3H3CCH3OHSOOOHH3CCH3CH3H3CCH3

CAS RN 15452-89-8

IUPAC Name: 2-[2-hydroxy-5-(2,4,4-trimethylpentan-2-yl)benzenesulfonyl]-4-(2,4,4-trimethylpentan-2-yl)phenol
Synonyms: 2-[2-hydroxy-5-(2,4,4-trimethylpentan-2-yl)benzenesulfonyl]-4-(2,4,4-trimethylpentan-2-yl)phenol
Molecular Weight (g/mol): 474.7
Molecular Formula: C28H42O4S
InChi Key: LMTGYJHIOQZSAA-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)C1=CC=C(O)C(=C1)S(=O)(=O)C1=CC(=CC=C1O)C(C)(C)CC(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results