accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSOOONa

CAS RN 15521-77-4

IUPAC Name: sodium pyridine-3-sulfonate
Synonyms: sodium pyridine-3-sulfonate
Molecular Weight (g/mol): 181.14
Molecular Formula: C5H4NNaO3S
InChi Key: CQSNULMCHIOCCX-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CN=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results