CAS RN 155270-99-8
IUPAC Name:
8-[(1E)-2-(3,4-dimethoxyphenyl)ethenyl]-1,3-diethyl-7-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms:
istradefylline
Molecular Weight (g/mol):
384.44
Molecular Formula:
C20H24N4O4
InChi Key:
IQVRBWUUXZMOPW-PKNBQFBNSA-N
SMILES:
CCN1C2=C(N(C)C(\C=C\C3=CC=C(OC)C(OC)=C3)=N2)C(=O)N(CC)C1=O
1
–
1
of
1
results
1
Tocris Bioscience™ Istradefylline
CAS: 155270-99-8 Molecular Formula: C20H24N4O4