accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNCSOClClHCl

CAS RN 155512-52-0

IUPAC Name: 2-(3,4-dichlorophenyl)-N-[1-(3-isothiocyanatophenyl)-2-(pyrrolidin-1-yl)ethyl]-N-methylacetamide hydrochloride
Synonyms: 2-(3,4-dichlorophenyl)-N-[1-(3-isothiocyanatophenyl)-2-(pyrrolidin-1-yl)ethyl]-N-methylacetamide hydrochloride
Molecular Weight (g/mol): 484.86
Molecular Formula: C22H24Cl3N3OS
InChi Key: BNWYENYHNOESCX-UHFFFAOYNA-N
SMILES: Cl.CN(C(CN1CCCC1)C1=CC(=CC=C1)N=C=S)C(=O)CC1=CC(Cl)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results