accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOOH3CH3CO

CAS RN 1563-66-2

IUPAC Name: 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl N-methylcarbamate
Synonyms: carbofuran furadan
Molecular Weight (g/mol): 221.26
Molecular Formula: C12H15NO3
InChi Key: DUEPRVBVGDRKAG-UHFFFAOYSA-N
SMILES: CNC(=O)OC1=CC=CC2=C1OC(C)(C)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results