CAS RN 156886-85-0
IUPAC Name:
3,7,11,15-tetraazaheptadecane tetrahydrochloride
Synonyms:
3,7,11,15-tetraazaheptadecane tetrahydrochloride
Molecular Weight (g/mol):
390.26
Molecular Formula:
C13H36Cl4N4
InChi Key:
LAWBSOKBIROCQP-UHFFFAOYSA-N
SMILES:
Cl.Cl.Cl.Cl.CCNCCCNCCCNCCCNCC
1
–
1
of
1
results
1
R&D Systems™ N1,N11-Diethylnorspermine tetrahydrochloride
CAS: 156886-85-0 Molecular Formula: C13H36Cl4N4