accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CH3COHOHO

CAS RN 15715-58-9

IUPAC Name: carbonic acid; triethylamine
Synonyms: carbonic acid; triethylamine
Molecular Weight (g/mol): 163.22
Molecular Formula: C7H17NO3
InChi Key: AFQIYTIJXGTIEY-UHFFFAOYSA-N
SMILES: OC(O)=O.CCN(CC)CC
For Use With (Equipment) 
  • (2)
Quantity 
  • (1)
  • (1)
pH 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results