accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3H3CNCH3OH3CH3C

CAS RN 158493-17-5

IUPAC Name: 1,4a,6a,10-tetramethyl-7-(6-methylheptan-2-yl)-hexadecahydro-1H-indeno[5,4-f]quinolin-2-one
Synonyms: 1,4a,6a,10-tetramethyl-7-(6-methylheptan-2-yl)-dodecahydro-3H-indeno[5,4-f]quinolin-2-one
Molecular Weight (g/mol): 415.71
Molecular Formula: C28H49NO
InChi Key: XUTZDXHKQDPUMA-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)C1CCC2C3C(C)CC4N(C)C(=O)CCC4(C)C3CCC12C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)