accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHNFSO

CAS RN 158848-32-9

IUPAC Name: 3-(2-{4-[(benzenesulfinyl)methyl]-4-methoxypiperidin-1-yl}ethyl)-5-fluoro-1H-indole
Synonyms: 3-(2-{4-[(benzenesulfinyl)methyl]-4-methoxypiperidin-1-yl}ethyl)-5-fluoro-1H-indole
Molecular Weight (g/mol): 414.54
Molecular Formula: C23H27FN2O2S
InChi Key: BANYJBHWTOJQDU-UHFFFAOYNA-N
SMILES: COC1(CS(=O)C2=CC=CC=C2)CCN(CCC2=CNC3=CC=C(F)C=C23)CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results