accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOOHNH2NH2HClHCl(E/Z)

CAS RN 159190-44-0

IUPAC Name: 2-amino-5-[(1-aminoethylidene)amino]pentanoic acid dihydrochloride
Synonyms: 2-amino-5-[(1-aminoethylidene)amino]pentanoic acid dihydrochloride
Molecular Weight (g/mol): 246.13
Molecular Formula: C7H17Cl2N3O2
InChi Key: RYCMAAFECCXGHI-UHFFFAOYNA-N
SMILES: Cl.Cl.CC(N)=NCCCC(N)C(O)=O
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results