CAS RN 1597402-27-1
CAS RN 1597402-27-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ KYT 0353
Potent and selective inhibitor of L-type amino acid transporter 1 (LAT1)