CAS RN 15982-84-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ BCI
DUSP6 and DUSP1 inhibitor
2
MilliporeSigma™ Calbiochem™ Dual Specificity Protein Phosphatase 1/6 Inhibitor, BCI
The Dual Specificity Protein Phosphatase 1/6 Inhibitor, BCI controls the biological activity of Dual Specificity Protein Phosphatase 1/6. Primarily used for Phosphorylation & Dephosphorylation apps.