CAS RN 160312-62-9
IUPAC Name:
2-amino-N-[7,7-dimethyl-1-({[4-(2-methylphenyl)piperazin-1-yl]sulfonyl}methyl)bicyclo[2.2.1]heptan-2-yl]-4-methanesulfonylbutanamide hydrochloride
Synonyms:
2-amino-N-(7,7-dimethyl-1-{[4-(2-methylphenyl)piperazin-1-ylsulfonyl]methyl}bicyclo[2.2.1]heptan-2-yl)-4-methanesulfonylbutanamide hydrochloride
Molecular Weight (g/mol):
591.22
Molecular Formula:
C26H43ClN4O5S2
InChi Key:
GIUFQWFJHXXXEQ-UHFFFAOYNA-N
SMILES:
Cl.CC1=CC=CC=C1N1CCN(CC1)S(=O)(=O)CC12CCC(CC1NC(=O)C(N)CCS(C)(=O)=O)C2(C)C
1
–
1
of
1
results
1
R&D Systems™ L-368,899 hydrochloride
CAS: 160312-62-9 Molecular Formula: C26H43ClN4O5S2