accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3N(E)

CAS RN 1608-82-8

IUPAC Name: (2E)-3-methoxy-2-(methoxymethyl)prop-2-enenitrile
Synonyms: (2E)-3-methoxy-2-(methoxymethyl)prop-2-enenitrile
Molecular Weight (g/mol): 127.14
Molecular Formula: C6H9NO2
InChi Key: QZFRWRSOUHEOFM-GQCTYLIASA-N
SMILES: COC\C(=C\OC)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results