accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 160842-62-6

CH3H3CCH3NNOClCH3

CAS RN 160842-62-6

IUPAC Name: 3-tert-butyl-1-methyl-1H-pyrazole-5-carbonyl chloride
Synonyms: 5-tert-butyl-2-methylpyrazole-3-carbonyl chloride
Molecular Weight (g/mol): 200.67
Molecular Formula: C9H13ClN2O
InChi Key: SJGURXGUNLVMAQ-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C(Cl)=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results