CAS RN 161552-03-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Cyclo(-RGDfK)
Potent integrin inhibitor (αvβ3/αvβ6); also improves stem cell attachment in 3D cell culture scaffolds