Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Cyclo(-RGDfK) is a cyclic peptide, potent integrin inhibitor (IC50 values are 2.3 and 55 nM for αvβ3 and αvβ6, respectively). Cyclo(-RGDfK) improves human pluripotent stem cell (hPSC) adhesion and proliferaton on 3D cell culture synthetic surfaces. Can also be used to target fluorophores and cargo to cancer cells as αvβ3 integrin is upregulated on cancer cells.
Specifications
Specifications
| CAS | 161552-03-0 |
| Quantity | 5 mg |
| Target | Cell Adhesion Inhibitors |
| Molecular Formula | C27H41N9O7 |
| Purity | 95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
