CAS RN 1619-62-1
CAS RN 1619-62-1
IUPAC Name:
                                            
                                                    1,3-diethyl 2,2-dimethylpropanedioate
                                            
                                        
                                    
                                    
                                    
                                    
                                        Synonyms:
                                        
                                                1,3-diethyl 2,2-dimethylpropanedioate
                                        
                                    
                                                Molecular Weight (g/mol):
                                                
                                                           188.22
                                                
                                            
                                            
                                                 Molecular Formula:
                                                
                                                        C9H16O4
                                            
                                                
                                            
                                        InChi Key:
                                                
                                                        UELKSYXXNGHTSE-UHFFFAOYSA-N
                                                
                                            
                                            
                                            SMILES:
                                                
                                                            CCOC(=O)C(C)(C)C(=O)OCC
                                                
                                            
                                        
                                        1
                                        –
                                        1
                                        of
                                        1
                                        results
                                    
                                
                                        1 
                                    
                            
                                    
                                                        Diethyl dimethylmalonate, 97%, Thermo Scientific Chemicals
                                                
                                                
                                                
                                                    
                                                
                                                    
                                                
                                                                        CAS: 1619-62-1 Molecular Formula: C9H16O4 Molecular Weight (g/mol): 188.22 MDL Number: MFCD00009128 InChI Key: UELKSYXXNGHTSE-UHFFFAOYSA-N Synonym: diethyl 2,2-dimethylmalonate,diethyl dimethylmalonate,propanedioic acid, dimethyl-, diethyl ester,diethyldimethylmalonate,diethyl dimethyl malonate,1,3-diethyl 2,2-dimethylpropanedioate,malonic acid, dimethyl-, diethyl ester,2,2-dimethyl-malonic acid diethyl ester,zlchem 328,pubchem16321 PubChem CID: 15382 IUPAC Name: diethyl 2,2-dimethylpropanedioate SMILES: CCOC(=O)C(C)(C)C(=O)OCC
