accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNNNNNCH3CH3HNSOOH3C

CAS RN 1619994-69-2

IUPAC Name: ethyl N-[6-(3-methanesulfonamido-4-methylphenyl)-3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-8-yl]carbamate
Synonyms: ethyl N-[6-(3-methanesulfonamido-4-methylphenyl)-3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-8-yl]carbamate
Molecular Weight (g/mol): 404.45
Molecular Formula: C17H20N6O4S
InChi Key: UYBRROMMFMPJAN-UHFFFAOYSA-N
SMILES: CCOC(=O)NC1=CC(=NN2C(C)=NN=C12)C1=CC(NS(C)(=O)=O)=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)