CAS RN 1621903-81-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1S)-3-(1,10-Phenanthrolin-2-yl)-2'-phenyl-[1,1'-binaphthalen]-2-ol 97.0+%, TCI America™
CAS: 1621903-81-8 Molecular Formula: C38H24N2O Synonym: (S)-BinThro