accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 162641-16-9

IUPAC Name: 4-(diphenylmethoxy)-1-[3-(2H-1,2,3,4-tetrazol-5-yl)propyl]piperidine
Synonyms: 4-(diphenylmethoxy)-1-[3-(2H-1,2,3,4-tetrazol-5-yl)propyl]piperidine
Molecular Weight (g/mol): 377.49
Molecular Formula: C22H27N5O
InChi Key: TZQGXAHOROZEKN-UHFFFAOYSA-N
SMILES: C(CN1CCC(CC1)OC(C1=CC=CC=C1)C1=CC=CC=C1)CC1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results