CAS RN 163042-96-4
IUPAC Name:
(2S,3S,4R,5R)-5-(2-chloro-6-{[(3-iodophenyl)methyl]amino}-9H-purin-9-yl)-3,4-dihydroxy-N-methyloxolane-2-carboxamide
Synonyms:
(2S,3S,4R,5R)-5-(2-chloro-6-{[(3-iodophenyl)methyl]amino}purin-9-yl)-3,4-dihydroxy-N-methyloxolane-2-carboxamide
Molecular Weight (g/mol):
544.73
Molecular Formula:
C18H18ClIN6O4
InChi Key:
IPSYPUKKXMNCNQ-PFHKOEEOSA-N
SMILES:
CNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NCC3=CC=CC(I)=C3)N=C(Cl)N=C12
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Adenosine A3 Receptor Agonist, 2-Cl-IB-MECA
A highly selective agonist of adenosine A3 receptor (Ki = 330 pM).
2
Tocris Bioscience™ 2-Cl-IB-MECA
CAS: 163042-96-4 Molecular Formula: C18H18ClIN6O4