accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONCH3(S)(R)HOH3CH3CON

CAS RN 163163-23-3

IUPAC Name: N-[(3R,4S)-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-2H-1-benzopyran-4-yl]-N-methylethane-1-sulfonamide
Synonyms: N-[(3R,4S)-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-1-benzopyran-4-yl]-N-methylethanesulfonamide
Molecular Weight (g/mol): 324.4
Molecular Formula: C15H20N2O4S
InChi Key: HVSJHHXUORMCGK-UONOGXRCSA-N
SMILES: CCS(=O)(=O)N(C)[C@@H]1[C@@H](O)C(C)(C)OC2=CC=C(C=C12)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results