Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Chromanol 293B
GREENER_CHOICESDP

Catalog No. 141250 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-125-0 50 mg
14-121-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-125-0 Supplier R&D Systems™ Supplier No. 1412/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. IKs blocker. Also blocks ICFTR

Blocker of the slow delayed rectifier K+ current (IKs) (IC50 = 1-10 μM). Also blocks the CFTR chloride current (ICFTR) (IC50 = 19 μM).

Enantiomer also available.

Chemical Identifiers

CAS 163163-23-3
Molecular Formula C15H20N2O4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 324.39
Purity >99%
Molecular Weight (g/mol) 324.395
Chemical Name or Material Chromanol 293B
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.