accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16331-46-7

IUPAC Name: 4-ethoxybenzoyl chloride
Synonyms: 4-ethoxybenzoyl chloride
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: XLWQUESMILVIPR-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)