CAS RN 164513-39-7
IUPAC Name:
5-bromo-2-methoxy-4-methylpyridine
Synonyms:
5-bromo-2-methoxy-4-methylpyridine
Molecular Weight (g/mol):
202.05
Molecular Formula:
C7H8BrNO
InChi Key:
HTBPXLJKMNBQMS-UHFFFAOYSA-N
SMILES:
COC1=CC(C)=C(Br)C=N1
1
–
1
of
1
results
1
5-Bromo-2-methoxy-4-methylpyridine 98.0+%, TCI America™
CAS: 164513-39-7 Molecular Formula: C7H8BrNO Molecular Weight (g/mol): 202.051 MDL Number: MFCD04039980 InChI Key: HTBPXLJKMNBQMS-UHFFFAOYSA-N PubChem CID: 4178002 IUPAC Name: 5-bromo-2-methoxy-4-methylpyridine SMILES: CC1=CC(=NC=C1Br)OC